Merck
  • Organization of cholinergic, catecholaminergic, serotonergic and orexinergic nuclei in three strepsirrhine primates: Galago demidoff, Perodicticus potto and Lemur catta.
Organization of cholinergic, catecholaminergic, serotonergic and orexinergic nuclei in three strepsirrhine primates: Galago demidoff, Perodicticus potto and Lemur catta.
Journal of chemical neuroanatomy (2015-11-13)
Tanya Calvey, Nina Patzke, Consolate Kaswera-Kyamakya, Emmanuel Gilissen, Mads F Bertelsen, John D Pettigrew, Paul R Manger
ABSTRACT
The nuclear organization of the cholinergic, catecholaminergic, serotonergic and orexinergic systems in the brains of three species of strepsirrhine primates is presented. We aimed to investigate the nuclear complement of these neural systems in comparison to those of simian primates, megachiropterans and other mammalian species. The brains were coronally sectioned and immunohistochemically stained with antibodies against choline acetyltransferase, tyrosine hydroxylase, serotonin and orexin-A. The nuclei identified were identical among the strepsirrhine species investigated and identical to previous reports in simian primates. Moreover, a general similarity to other mammals was found, but specific differences in the nuclear complement highlighted potential phylogenetic interrelationships. The central feature of interest was the structure of the locus coeruleus complex in the primates, where a central compactly packed core (A6c) of tyrosine hydroxylase immunopositive neurons was surrounded by a shell of less densely packed (A6d) tyrosine hydroxylase immunopositive neurons. This combination of compact and diffuse divisions of the locus coeruleus complex is only found in primates and megachiropterans of all the mammalian species studied to date. This neural character, along with variances in a range of other neural characters, supports the phylogenetic grouping of primates with megachiropterans as a sister group.
Sigma-Aldrich
Anti-Orexin-A Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat