Merck
  • Leptin promotes methionine adenosyltransferase 2A expression in hepatic stellate cells by the downregulation of E2F-4 via the β-catenin pathway.
Leptin promotes methionine adenosyltransferase 2A expression in hepatic stellate cells by the downregulation of E2F-4 via the β-catenin pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-02-29)
Fangyun Cheng, Shengyan Su, Xiaofei Zhu, Xin Jia, Haimeng Tian, Xuguang Zhai, Wei Guan, Yajun Zhou
ABSTRACT
Most obese patients develop hyperleptinaemia. Leptin, mainly produced by adipocytes, demonstrates a promotional role in liver fibrosis. Hepatic stellate cell (HSC) activation, a key step in liver fibrogenesis, requires global reprogramming of gene expression. The remodeling of DNA methylation is a mechanism of the epigenetic regulation of gene expression. The biosynthesis of S-adenosylmethionine, a principle biological methyl donor, is catalyzed by methionine adenosyltransferase (MAT) such as MATⅡ which has been shown to promote HSC activation in vitro. This study was mainly aimed to determine the effect of leptin on MAT2A expression (the catalytic subunit of MATⅡ) in HSCs. Results showed that MAT2A knockdown reduced leptin-induced HSC activation and liver fibrosis in the leptin-deficient mouse model. Leptin promoted MAT2A expression in HSCs and increased MAT2A promoter activity. The axis of the β-catenin pathway/E2F-4 mediated the effect of leptin on MAT2A expression. Leptin-induced β-catenin signaling reduced E2F-4 expression and thus abated E2F-4 binding to MAT2A promoter at a site around -2779 bp, leading to an increase in the MAT2A promoter activity. These data might shed more light on the mechanisms responsible for liver fibrogenesis in obese patients with hyperleptinaemia.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAT2A
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
Germane, electronic grade, ≥99.997%