Merck
  • Infant High-Grade Gliomas Comprise Multiple Subgroups Characterized by Novel Targetable Gene Fusions and Favorable Outcomes.
Infant High-Grade Gliomas Comprise Multiple Subgroups Characterized by Novel Targetable Gene Fusions and Favorable Outcomes.
Cancer discovery (2020-04-03)
Matthew Clarke, Alan Mackay, Britta Ismer, Jessica C Pickles, Ruth G Tatevossian, Scott Newman, Tejus A Bale, Iris Stoler, Elisa Izquierdo, Sara Temelso, Diana M Carvalho, Valeria Molinari, Anna Burford, Louise Howell, Alex Virasami, Amy R Fairchild, Aimee Avery, Jane Chalker, Mark Kristiansen, Kelly Haupfear, James D Dalton, Wilda Orisme, Ji Wen, Michael Hubank, Kathreena M Kurian, Catherine Rowe, Mellissa Maybury, Stephen Crosier, Jeffrey Knipstein, Ulrich Schüller, Uwe Kordes, David E Kram, Matija Snuderl, Leslie Bridges, Andrew J Martin, Lawrence J Doey, Safa Al-Sarraj, Christopher Chandler, Bassel Zebian, Claire Cairns, Rachael Natrajan, Jessica K R Boult, Simon P Robinson, Martin Sill, Ira J Dunkel, Stephen W Gilheeney, Marc K Rosenblum, Debbie Hughes, Paula Z Proszek, Tobey J Macdonald, Matthias Preusser, Christine Haberler, Irene Slavc, Roger Packer, Ho-Keung Ng, Shani Caspi, Mara Popović, Barbara Faganel Kotnik, Matthew D Wood, Lissa Baird, Monika Ashok Davare, David A Solomon, Thale Kristin Olsen, Petter Brandal, Michael Farrell, Jane B Cryan, Michael Capra, Michael Karremann, Jens Schittenhelm, Martin U Schuhmann, Martin Ebinger, Winand N M Dinjens, Kornelius Kerl, Simone Hettmer, Torsten Pietsch, Felipe Andreiuolo, Pablo Hernáiz Driever, Andrey Korshunov, Lotte Hiddingh, Barbara C Worst, Dominik Sturm, Marc Zuckermann, Olaf Witt, Tabitha Bloom, Clare Mitchell, Evelina Miele, Giovanna Stefania Colafati, Francesca Diomedi-Camassei, Simon Bailey, Andrew S Moore, Timothy E G Hassall, Stephen P Lowis, Maria Tsoli, Mark J Cowley, David S Ziegler, Matthias A Karajannis, Kristian Aquilina, Darren R Hargrave, Fernando Carceller, Lynley V Marshall
ABSTRACT
Infant high-grade gliomas appear clinically distinct from their counterparts in older children, indicating that histopathologic grading may not accurately reflect the biology of these tumors. We have collected 241 cases under 4 years of age, and carried out histologic review, methylation profiling, and custom panel, genome, or exome sequencing. After excluding tumors representing other established entities or subgroups, we identified 130 cases to be part of an "intrinsic" spectrum of disease specific to the infant population. These included those with targetable MAPK alterations, and a large proportion of remaining cases harboring gene fusions targeting ALK (n = 31), NTRK1/2/3 (n = 21), ROS1 (n = 9), and MET (n = 4) as their driving alterations, with evidence of efficacy of targeted agents in the clinic. These data strongly support the concept that infant gliomas require a change in diagnostic practice and management. SIGNIFICANCE: Infant high-grade gliomas in the cerebral hemispheres comprise novel subgroups, with a prevalence of ALK, NTRK1/2/3, ROS1, or MET gene fusions. Kinase fusion-positive tumors have better outcome and respond to targeted therapy clinically. Other subgroups have poor outcome, with fusion-negative cases possibly representing an epigenetically driven pluripotent stem cell phenotype.See related commentary by Szulzewsky and Cimino, p. 904.This article is highlighted in the In This Issue feature, p. 890.
Sigma-Aldrich
Germane, electronic grade, ≥99.997%
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 29.04 mPa.s (25 °C)