Merck
  • Synthesis, characterization and antioxidant activity of chitosan Schiff base derivatives bearing (-)-gossypol.
Synthesis, characterization and antioxidant activity of chitosan Schiff base derivatives bearing (-)-gossypol.
Carbohydrate polymers (2020-06-02)
Neslihan Beyazit, Halide Sinem Çakran, Ali Cabir, Yaşar Akışcan, Cahit Demetgül
ABSTRACT
In this work, two new chitosan-Schiff base derivatives (HCS-GSP and LCS-GSP) were synthesized by the condensation reaction of high molecular weight chitosan (HCS) and low molecular weight chitosan (LCS) with (-)-gossypol (GSP), respectively. For this purpose, racemic gossypol was isolated from cotton seeds and it was further enantiomerically purified by diastereomeric resolution technique using l-tryptophan methyl ester hydrochloride. Then, chitosan polymers were derivatized with (-)-gossypol by the condensation reaction. The isolated and synthesized coumpounds were characterized by physical measurements and spectroscopic methods (elemental analysis C,H,N, Uv-vis, FT-IR, 1H&13C NMR and TG/DTG/DTA). The antioxidant activity of high molecular weight chitosan (HCS), low molecular weight chitosan (LCS) and their gossypol derivatives was evaluated as radical scavengers against 1,1-diphenyl-2-picrylhydrazyl radicals (DPPH). The results showed that both of the chitosan-gossypol derivatives (HCS-GSP and LCS-GSP) had a better ability to scavenging DPPH radical (IC50, 12 μg/mL and 16 μg/mL, respectively) than its unmodified chitosan.
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%
Sigma-Aldrich
Chitosan, low molecular weight