Merck
  • Ligand-enabled cross-coupling of C(sp3)-H bonds with arylboron reagents via Pd(II)/Pd(0) catalysis.
Ligand-enabled cross-coupling of C(sp3)-H bonds with arylboron reagents via Pd(II)/Pd(0) catalysis.
Nature chemistry (2014-01-24)
Kelvin S L Chan, Masayuki Wasa, Ling Chu, Brian N Laforteza, Masanori Miura, Jin-Quan Yu
ABSTRACT
There have been numerous developments in C-H activation reactions in the past decade. Attracted by the ability to functionalize molecules directly at ostensibly unreactive C-H bonds, chemists have discovered reaction conditions that enable reactions of C(sp(2))-H and C(sp(3))-H bonds with a variety of coupling partners. Despite these advances, the development of suitable ligands that enable catalytic C(sp(3))-H bond functionalization remains a significant challenge. Herein we report the discovery of a mono-N-protected amino acid ligand that enables Pd(II)-catalysed coupling of Ī³-C(sp(3))-H bonds in triflyl-protected amines with arylboron reagents. Remarkably, no background reaction was observed in the absence of ligand. A variety of amine substrates and arylboron reagents were cross-coupled using this method. Arylation of optically active substrates derived from amino acids also provides a potential route for preparing non-proteinogenic amino acids.
Sigma-Aldrich
4-Methoxycarbonylphenylboronic acid pinacol ester, 97%