Merck
  • Amyloid β Is Not the Major Factor Accounting for Impaired Adult Hippocampal Neurogenesis in Mice Overexpressing Amyloid Precursor Protein.
Amyloid β Is Not the Major Factor Accounting for Impaired Adult Hippocampal Neurogenesis in Mice Overexpressing Amyloid Precursor Protein.
Stem cell reports (2016-10-04)
Hongyu Pan, Dongpi Wang, Xiaoqin Zhang, Dongming Zhou, Heng Zhang, Qi Qian, Xiao He, Zhaoling Liu, Yunjin Liu, Tingting Zheng, Ling Zhang, Mingkai Wang, Binggui Sun
ABSTRACT
Adult hippocampal neurogenesis was impaired in several Alzheimer's disease models overexpressing mutant human amyloid precursor protein (hAPP). However, the effects of wild-type hAPP on adult neurogenesis and whether the impaired adult hippocampal neurogenesis was caused by amyloid β (Aβ) or APP remained unclear. Here, we found that neurogenesis was impaired in the dentate gyrus (DG) of adult mice overexpressing wild-type hAPP (hAPP-I5) compared with controls. However, the adult hippocampal neurogenesis was more severely impaired in hAPP-I5 than that in hAPP-J20 mice, which express similar levels of hAPP mRNA but much higher levels of Aβ. Furthermore, reducing Aβ levels did not affect the number of doublecortin-positive cells in the DG of hAPP-J20 mice. Our results suggested that hAPP was more likely an important factor inhibiting adult neurogenesis, and Aβ was not the major factor affecting neurogenesis in the adult hippocampus of hAPP mice.
Sigma-Aldrich
Anti-NeuN (rabbit monoclonal) Antibody, clone 27-4, clone 27-4, from rabbit
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®
Roche
Anti-Bromodeoxyuridine, from mouse IgG1 (clone: BMC9318)