Merck
  • Trafficking of multipotent mesenchymal stromal cells from maternal circulation through the placenta involves vascular endothelial growth factor receptor-1 and integrins.
Trafficking of multipotent mesenchymal stromal cells from maternal circulation through the placenta involves vascular endothelial growth factor receptor-1 and integrins.
Stem cells (Dayton, Ohio) (2007-11-03)
Chie-Pein Chen, Ming-Yi Lee, Jian-Pei Huang, John D Aplin, Yi-Hsin Wu, Cing-Siang Hu, Pei-Chun Chen, Hung Li, Shiaw-Min Hwang, Shu-Hsiang Liu, Yuh-Cheng Yang
ABSTRACT
Maternal cells can become engrafted in various fetal organs during pregnancy. The nature of the cells and the mechanisms of maternofetal cell trafficking are not clear. We demonstrate that human lineage-negative, CD34-negative (Lin(-)CD34(-)) multipotent mesenchymal stromal cells express alpha(2), alpha(4), alpha(5), and beta(1) integrins, which mediate their adhesion to endothelium, and vascular endothelial growth factor receptor-1 (VEGFR-1), which mediates their response to vascular endothelial growth factor A (VEGF-A). A maternal-fetal VEGF-A concentration gradient exists across the placental barrier, and cord blood plasma induces transendothelial and trans-Matrigel migration of stem cells in vitro. Migration is inhibited by a VEGF-A-neutralizing antibody or antibodies against VEGFR-1 or integrin alpha(2), alpha(4), alpha(5), or beta(1). When Lin(-)CD34(-) multipotent mesenchymal stromal cells are transferred to rat maternal venous blood, they traffic through the placenta, engraft in various fetal organs, and persist in offspring for at least 12 weeks. Cell proliferation ability is retained in the xenogeneic placenta. Maternofetal trafficking is significantly reduced by blocking antibodies against integrins alpha(2), alpha(4), alpha(5), and beta(1) or VEGFR-1. These results suggest that maternal microchimerism arises by the trafficking of multipotent mesenchymal stromal cells via VEGF-A- and integrin-dependent pathways across the hemochorial placenta to fetal tissues.
Sigma-Aldrich
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, clone KDR-1, ascites fluid
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, a.a. 82-87, clone JB1A (a.k.a. J10), ascites fluid, clone JB1A (J10), Chemicon®