Merck
  • Evaluation of the Therapeutic Potential of the Novel Isotype Specific HDAC Inhibitor 4SC-202 in Urothelial Carcinoma Cell Lines.
Evaluation of the Therapeutic Potential of the Novel Isotype Specific HDAC Inhibitor 4SC-202 in Urothelial Carcinoma Cell Lines.
Targeted oncology (2016-06-03)
Maria Pinkerneil, Michèle J Hoffmann, Hella Kohlhof, Wolfgang A Schulz, Günter Niegisch
ABSTRACT
Targeting of class I histone deacetylases (HDACs) exerts antineoplastic actions in various cancer types by modulation of transcription, upregulation of tumor suppressors, induction of cell cycle arrest, replication stress and promotion of apoptosis. Class I HDACs are often deregulated in urothelial cancer. 4SC-202, a novel oral benzamide type HDAC inhibitor (HDACi) specific for class I HDACs HDAC1, HDAC2 and HDAC3 and the histone demethylase LSD1, shows substantial anti-tumor activity in a broad range of cancer cell lines and xenograft tumor models. The aim of this study was to investigate the therapeutic potential of 4SC-202 in urothelial carcinoma (UC) cell lines. We determined dose response curves of 4SC-202 by MTT assay in seven UC cell lines with distinct HDAC1, HDAC2 and HDAC3 expression profiles. Cellular effects were further analyzed in VM-CUB1 and UM-UC-3 cells by colony forming assay, caspase-3/7 assay, flow cytometry, senescence assay, LDH release assay, and immunofluorescence staining. Response markers were followed by quantitative real-time PCR and western blotting. Treatment with the class I HDAC specific inhibitor SAHA (vorinostat) served as a general control. 4SC-202 significantly reduced proliferation of all epithelial and mesenchymal UC cell lines (IC50 0.15-0.51 μM), inhibited clonogenic growth and induced caspase activity. Flow cytometry revealed increased G2/M and subG1 fractions in VM-CUB1 and UM-UC-3 cells. Both effects were stronger than with SAHA treatment. Specific pharmacological inhibition of class I HDACs by 4SC-202 impairs UC cell viability, inducing cell cycle disturbances and cell death. Combined inhibition of HDAC1, HDAC2 and HDAC3 seems to be a promising treatment strategy for UC.
Sigma-Aldrich
Q-VD-OPh hydrate, ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP18, ascites fluid, clone BP18, Upstate®