Merck
  • Long-term in vivo imaging of fibrillar tau in the retina of P301S transgenic mice.
Long-term in vivo imaging of fibrillar tau in the retina of P301S transgenic mice.
PloS one (2013-01-10)
Christian Schön, Nadine A Hoffmann, Simon M Ochs, Steffen Burgold, Severin Filser, Sonja Steinbach, Mathias W Seeliger, Thomas Arzberger, Michel Goedert, Hans A Kretzschmar, Boris Schmidt, Jochen Herms
ABSTRACT
Tauopathies are widespread neurodegenerative disorders characterised by the intracellular accumulation of hyperphosphorylated tau. Especially in Alzheimer's disease, pathological alterations in the retina are discussed as potential biomarkers to improve early diagnosis of the disease. Using mice expressing human mutant P301S tau, we demonstrate for the first time a straightforward optical approach for the in vivo detection of fibrillar tau in the retina. Longitudinal examinations of individual animals revealed the fate of single cells containing fibrillar tau and the progression of tau pathology over several months. This technique is most suitable to monitor therapeutic interventions aimed at reducing the accumulation of fibrillar tau. In order to evaluate if this approach can be translated to human diagnosis, we tried to detect fibrillar protein aggregates in the post-mortem retinas of patients that had suffered from Alzheimer's disease or Progressive Supranuclear Palsy. Even though we could detect hyperphosphorylated tau, we did not observe any fibrillar tau or Aß aggregates. In contradiction to previous studies, our observations do not support the notion that Aβ or tau in the retina are of diagnostic value in Alzheimer's disease.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Albumin, Mouse