Merck
  • Cigarette smoke metal-catalyzed protein oxidation leads to vascular endothelial cell contraction by depolymerization of microtubules.
Cigarette smoke metal-catalyzed protein oxidation leads to vascular endothelial cell contraction by depolymerization of microtubules.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2005-06-30)
David Bernhard, Adam Csordas, Blair Henderson, Andrea Rossmann, Michaela Kind, Georg Wick
ABSTRACT
Smoking is a significant risk factor for development of atherosclerosis. However, the pathophysiology of smoking-mediated vessel wall damage is not understood. With tools ranging from analytical chemistry to cell biology, we show that cigarette smoke contains metals that catalyze the direct oxidation of cellular proteins by smoke oxidants. Oxidation of cellular proteins causes a loss of microtubule function, culminating in microtubule depolymerization and proteasome-dependent degradation of alpha-tubulin. As a consequence of the microtubule collapse, cytoskeletal structures as well as intermediate filaments break down, leading finally to a contraction of vascular endothelial cells. We observed a smoke extract-induced, calpain-dependent degradation of the intracellular form of platelet-endothelial cell adhesion molecule 1/CD31, as well as a release of P-selectin/CD62P, IL-6, and IL-8 from endothelial cells into the supernatant. Increased levels of soluble CD62P and IL-6 are well known to be associated with smoking in humans. Increased permeability of the vascular endothelium is a crucial event in atherogenesis. This work highlights the compounds and mechanisms by which cigarette smoke induces leakiness of the vascular endothelium.
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.