Merck
  • KDM6B Counteracts EZH2-Mediated Suppression of IGFBP5 to Confer Resistance to PI3K/AKT Inhibitor Treatment in Breast Cancer.
KDM6B Counteracts EZH2-Mediated Suppression of IGFBP5 to Confer Resistance to PI3K/AKT Inhibitor Treatment in Breast Cancer.
Molecular cancer therapeutics (2018-06-22)
Wenyu Wang, Keng Gat Lim, Min Feng, Yi Bao, Puay Leng Lee, Yu Cai, Yufeng Chen, Hao Zhang, Diego Marzese, Dave S B Hoon, Qiang Yu
ABSTRACT
Despite showing promise against PIK3CA-mutant breast cancers in preclinical studies, PI3K/AKT pathway inhibitors demonstrate limited clinical efficacy as monotherapy. Here, we found that histone H3K27me3 demethylase KDM6B-targeted IGFBP5 expression provides a protective mechanism for PI3K/AKT inhibitor-induced apoptosis in breast cancer cells. We found that overexpression of KDM6B and IGFBP5 in luminal breast cancer are positively associated with poorer disease outcomes. Mechanistically, KDM6B promotes IGFBP5 expression by antagonizing EZH2-mediated repression, and pharmacologic inhibition of KDM6B augments apoptotic response to PI3K/AKT inhibitor treatment. Moreover, the IGFBP5 expression is upregulated upon acquired resistance to the PI3K inhibitor GDC-0941, which is associated with an epigenetic switch from H3K27me3 to H3K27Ac at the IGFBP5 gene promoter. Intriguingly, GDC-0941-resistant breast cancer cells remained sensitive to KDM6B or IGFBP5 inhibition, indicating the dependency on the KDM6B-IGFBP5 axis to confer the survival advantage in GDC-0941-resistant cells. Our study reveals an epigenetic mechanism associated with resistance to targeted therapy and demonstrates that therapeutic targeting of KDM6B-mediated IGFBP5 expression may provide a useful approach to mitigate both intrinsic and acquired resistance to the PI3K inhibitor in breast cancer. Mol Cancer Ther; 17(9); 1973-83. ©2018 AACR.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-UTX Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP5
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2