Merck
  • Lipopolysaccharide increases the incidence of collagen-induced arthritis in mice through induction of protease HTRA-1 expression.
Lipopolysaccharide increases the incidence of collagen-induced arthritis in mice through induction of protease HTRA-1 expression.
Arthritis and rheumatism (2013-08-29)
Yuzhu Hou, Haijiang Lin, Linnan Zhu, Zhaoting Liu, Fanlei Hu, Jianfeng Shi, Tao Yang, Xiaoyun Shi, Mingzhao Zhu, Bernard F Godley, Qiang Wang, Zhanguo Li, Yong Zhao
ABSTRACT
The protease HTRA-1 is closely associated with rheumatoid arthritis (RA). The molecular mechanisms that control HTRA-1 expression are currently unknown. This study was undertaken to determine the regulatory role of Toll-like receptors (TLRs) on HTRA-1 expression in mice with collagen-induced arthritis (CIA) and in synovial cells from RA patients. HTRA-1 messenger RNA and protein production in mouse fibroblasts, mouse macrophages, and freshly isolated RA patient synovial cells treated with TLR ligands were detected by real-time polymerase chain reaction and enzyme-linked immunosorbent assay, respectively. Arthritis incidence and severity were determined using clinical scores and histopathologic analysis. Involvement of HTRA-1 in lipopolysaccharide (LPS)-increased arthritis incidence and severity in mice was determined using anti-HTRA-1 monoclonal antibody. The signal pathways involved in HTRA-1 expression were accessed by specific inhibitors, RNA interference, dual-luciferase reporter, and chromatin immunoprecipitation methods. LPS and tenascin-C, but not the other TLR ligands tested, strongly induced HTRA-1 expression. LPS significantly increased HTRA-1 expression in the joint tissue as well as arthritis incidence and severity in mice with CIA. Blocking HTRA-1 by antibody significantly decreased LPS-promoted CIA severity. Inhibiting NF-ĪŗB significantly decreased LPS-induced HTRA-1 expression in mouse and human cells. Dual-luciferase reporter assay and ChIP analysis showed that p65 directly binds to HTRA-1 promoter (amino acid 347). Our findings indicate that TLR-4 activation increases HTRA-1 expression through the NF-ĪŗB pathway in fibroblasts and macrophages. HTRA-1 expression is involved in the enhancing effects of LPS on CIA. This study offers new insights into the regulation of HTRA-1 expression via LPS/TLR-4 and the role of HTRA-1 in RA pathogenesis.
Sigma-Aldrich
Anti-HtrA1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution