
Valacyclovir Related Compound E

Synonym(s):
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-(benzyloxycarbonyl)-L-valinate, N-[(Phenylmethoxy)carbonyl]-L-valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester
Empirical Formula (Hill Notation):
C21H26N6O6
CAS Number:
Molecular Weight:
458.47
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing