
Tropicamide Related Compound D

Synonym(s):
Phenylacetic acid, α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA
Linear Formula:
C6H5CH2CO2H
CAS Number:
Molecular Weight:
136.15
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-2 of 2