
Tris(2-carboxyethyl)phosphine hydrochloride

Synonym(s):
TCEP
Empirical Formula (Hill Notation):
C9H15O6P · HCl
CAS Number:
Molecular Weight:
286.65
Products
Product Category
Brand
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Technique