Thionin acetate salt
Synonym(s):
3,7-Diamino-5-phenothiazinium acetate, Lauth’s violet
Empirical Formula (Hill Notation):
C12H9N3S · C2H4O2
CAS Number:
Molecular Weight:
287.34
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing