Skip to Content
Merck
tert-Butyldimethylsilyl chloride reagent grade, 97%

tert-Butyldimethylsilyl chloride

tert-Butyldimethylsilyl chloride reagent grade, 97%
Synonym(s):
tert-Butyl(chloro)dimethylsilane, tert-Butyldimethylchlorosilane, TBDMSCl
Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing