Sodium nitroprusside dihydrate
Synonym(s):
Nitroprusside sodium, SNP, Sodium nitroferricyanide, Sodium pentacyanonitrosylferrate
Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
Products
Product Category
Formula Weight
Physical Form
Purity
Agency
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing