![Sodium bis(trimethylsilyl)amide solution 0.6 M in toluene](/deepweb/assets/sigmaaldrich/product/structures/361/254/1cd613aa-a904-43a0-ab4e-7fa976f763e4/640/1cd613aa-a904-43a0-ab4e-7fa976f763e4.png)
Sodium bis(trimethylsilyl)amide solution
![Sodium bis(trimethylsilyl)amide solution 0.6 M in toluene](/deepweb/assets/sigmaaldrich/product/structures/361/254/1cd613aa-a904-43a0-ab4e-7fa976f763e4/640/1cd613aa-a904-43a0-ab4e-7fa976f763e4.png)
Synonym(s):
Hexamethyldisilazane sodium salt solution
Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-3 of 3