
Sodium 1-nonanesulfonate

Synonym(s):
1-Nonanesulfonic acid sodium salt
Linear Formula:
CH3(CH2)8SO3Na
CAS Number:
Molecular Weight:
230.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing