
Samarium

Synonym(s):
Samarium
Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Element
Quality Segment
Showing 1-15 of 15
Compare
Product Number
Description
Pricing