Robenidine hydrochloride
Synonym(s):
1,3-Bis[(4-chlorobenzylidene)amino]guanidine monohydrochloride
Empirical Formula (Hill Notation):
C15H13Cl2N5 · HCl
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing