Skip to Content
Merck
vibrant-m

Ribavirin Related Compound D

vibrant-m
Synonym(s):
1H-1,2,4-Triazole-3-carboxamide
Empirical Formula (Hill Notation):
C3H4N4O
CAS Number:
Molecular Weight:
112.09

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade