
Repaglinide Related Compound A

Synonym(s):
(S)-3-Methyl-1-[2-(1-piperidinyl)phenyl]butylamine N-acetyl-L-glutamate salt
Empirical Formula (Hill Notation):
C16H26N2 · C7H11NO5
CAS Number:
Molecular Weight:
435.56
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2