![Pyrvinium pamoate salt hydrate ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/292/359/5a9788a7-dad3-4e88-b3e5-e2d5911ec35f/640/5a9788a7-dad3-4e88-b3e5-e2d5911ec35f.png)
Pyrvinium pamoate salt hydrate
![Pyrvinium pamoate salt hydrate ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/292/359/5a9788a7-dad3-4e88-b3e5-e2d5911ec35f/640/5a9788a7-dad3-4e88-b3e5-e2d5911ec35f.png)
Synonym(s):
6-(Dimethylamino)-2-[2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethenyl]-1-methyl-4,4′-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (2:1)-quinolinium, Alnoxin, Altolat, NSC 223622, PP, Pamovin, Vermitibier, Vipyrvinium embonate
Empirical Formula (Hill Notation):
C26H28N3 · 0.5 C23H14O6 · xH2O
CAS Number:
Molecular Weight:
575.70 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing