
Pyrazine

Synonym(s):
Pyrazine
Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing