Propofol Related Compound A
Synonym(s):
3,3′-5,5′-Tetraisopropyldiphenol, Dipropofol
Empirical Formula (Hill Notation):
C24H34O2
CAS Number:
Molecular Weight:
354.53
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing