
p-Phenylenediamine

Synonym(s):
1,4-Benzenediamine, 1,4-Diaminobenzene, 1,4-Phenylenediamine
Linear Formula:
C6H4(NH2)2
CAS Number:
Molecular Weight:
108.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing