
Plastic additive 14

Synonym(s):
3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Empirical Formula (Hill Notation):
C41H82O6P2
CAS Number:
Molecular Weight:
733.03
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing