Skip to Content
Merck
Oxycodone certified reference material, pharmaceutical secondary standard

Oxycodone

Oxycodone certified reference material, pharmaceutical secondary standard
Synonym(s):
(5α)-4,5-Epoxy-14-hydroxy-3-methoxy-17-methylmorphinan-6-one, 14-Hydroxydihydrocodeinone, Dihydrone
Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36

Products

Product Category
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing