
Omeprazole for peak identification

Synonym(s):
Omeprazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Antra, Losec
Empirical Formula (Hill Notation):
C17H19N3O3S
CAS Number:
Molecular Weight:
345.42
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing