
Nutrient Mixture F-12 Ham

Synonym(s):
Ham’s F-12, Nutrient Mixture F-12 Ham
Products
Product Category
Brand
Physical Form
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing