![N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for GC derivatization, LiChropur™, contains 10% TMCS, 98% (excluding TMCS)](/deepweb/assets/sigmaaldrich/product/structures/149/727/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d/640/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d.png)
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane
![N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for GC derivatization, LiChropur™, contains 10% TMCS, 98% (excluding TMCS)](/deepweb/assets/sigmaaldrich/product/structures/149/727/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d/640/f39401aa-aa5c-4ef2-a3de-cf60c4164f2d.png)
Synonym(s):
BSTFA + TMCS
Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40
Products
Brand
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing