
N-Nitrosodibutylamine (NDBA)

Synonym(s):
N-Butyl-N-nitroso-1-butanamine
Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2