
Nivalenol solution

Synonym(s):
3α,4β,7α, 15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one, NIV
Empirical Formula (Hill Notation):
C15H20O7
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing