![n-Butyllithium solution 11.0 M in hexanes](/deepweb/assets/sigmaaldrich/product/structures/134/313/58c7cde4-ea5d-4ffd-a37e-f60648ab5079/640/58c7cde4-ea5d-4ffd-a37e-f60648ab5079.png)
n-Butyllithium solution
![n-Butyllithium solution 11.0 M in hexanes](/deepweb/assets/sigmaaldrich/product/structures/134/313/58c7cde4-ea5d-4ffd-a37e-f60648ab5079/640/58c7cde4-ea5d-4ffd-a37e-f60648ab5079.png)
Synonym(s):
n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide
Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group