![Monoclonal Anti-Vimentin antibody produced in mouse clone VI-01](/deepweb/assets/sigmaaldrich/product/images/408/423/81b2c808-bc9d-4d61-b11c-b9b10e5422ba/640/81b2c808-bc9d-4d61-b11c-b9b10e5422ba.jpg)
Monoclonal Anti-Vimentin antibody produced in mouse
![Monoclonal Anti-Vimentin antibody produced in mouse clone VI-01](/deepweb/assets/sigmaaldrich/product/images/408/423/81b2c808-bc9d-4d61-b11c-b9b10e5422ba/640/81b2c808-bc9d-4d61-b11c-b9b10e5422ba.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing