Skip to Content
Merck
Molecular sieves, 4 Å pellets, 3.2 mm diameter

Molecular sieves, 4 Å

Molecular sieves, 4 Å pellets, 3.2 mm diameter
Synonym(s):
Sodium Alumino-Silicate
Linear Formula:
Na12[(AlO2)12(SiO2)12] · xH2O
CAS Number:

Products

Product Category
Brand
Physical Form
Quality Segment
Pore Size (Å)
Showing 1-5 of 5