Skip to Content
Merck
Methyl cinnamate ≥99.0% (GC)

Methyl cinnamate

Methyl cinnamate ≥99.0% (GC)
Linear Formula:
C6H5CH=CHCOOCH3
CAS Number:
Molecular Weight:
162.19
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing