![Mesalazine British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/744/417af975-e5a9-4e46-bba5-b1fb5eda8a33/640/417af975-e5a9-4e46-bba5-b1fb5eda8a33.png)
Mesalazine
![Mesalazine British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/744/417af975-e5a9-4e46-bba5-b1fb5eda8a33/640/417af975-e5a9-4e46-bba5-b1fb5eda8a33.png)
Synonym(s):
5-Aminosalicylic acid, 5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine
Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing