Martin Sulfurane Dehydrating agent
Synonym(s):
Bis[α,α-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, Bis[α,α-bis(trifluoromethyl)benzyloxy]diphenylsulfur
Linear Formula:
[C6H5C(CF3)2O]2S(C6H5)2
CAS Number:
Molecular Weight:
672.52
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1