
Magnesium L-lactate hydrate

Synonym(s):
L-Lactic acid magnesium salt
Linear Formula:
[CH3CH(OH)COO]2Mg · xH2O
CAS Number:
Molecular Weight:
202.45 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Technique