
Lysophosphatidylcholine

Synonym(s):
L-α-Lysophosphatidylcholine from egg yolk, 1-Acyl-sn-glycero-3-phosphocholine, L-α-Lysolecithin
CAS Number:
Products
Brand
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2