
L-Threonine-13C4,15N

Linear Formula:
13CH313CH(OH)13CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
124.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing