Skip to Content
Merck
Leflunomide European Pharmacopoeia (EP) Reference Standard

Leflunomide

Leflunomide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
5-Methylisoxazole-4-(4-trifluoromethyl)carboxanilide, Leflunomide
Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique