
Jujuboside A

Synonym(s):
(3β,16β,23R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→2)-O0[O-β-D-glucopyranosyl-(1→6)-O-[β-D-xylopyranosyl-(1→2)-β-D-glucopyranosyl-(1→3)]-α-L-arabinopyranoside
Empirical Formula (Hill Notation):
C58H94O26
CAS Number:
Molecular Weight:
1207.35
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing