
Isopropylmagnesium bromide solution

Linear Formula:
(CH3)2MgBr
CAS Number:
Molecular Weight:
147.30
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing