
Iodipamide

Synonym(s):
3,3′(Adipoyldiimino)bis(2,4,6-triiodobenzoic acid), Adipiodone, Iodipamide
Empirical Formula (Hill Notation):
C20H14I6N2O6
CAS Number:
Molecular Weight:
1139.76
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing