
Indole

Synonym(s):
1H-Benzo[b]pyrrole, Indole
Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade