Skip to Content
Merck
Hexadecyltrimethylammonium bisulfate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)

Hexadecyltrimethylammonium bisulfate

Hexadecyltrimethylammonium bisulfate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)
Synonym(s):
Hexadecyltrimethylammonium hydrogen sulfate
Linear Formula:
CH3(CH2)15N(HSO4)(CH3)3
CAS Number:
Molecular Weight:
381.61
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1